Learn More
Chemglass Life Sciences Distilling Head, Vigreux, Jacketed, 10/18 Thermometer Joint, 14/20 Joint Size, 2mm Glass Stopcock, Approx. 255mm Height x 310mm Width
SDP
Catalog No. CG124708
Encompass_Preferred
Distilling Head, Vigreux, Jacketed, 10/18 Thermometer Joint,14/20 Joint Size, 2mm Glass Stopcock, Approx. 255mm Height x 310mm Width. Jacketed fractionation distilling head for use under vacuum. The 10/18 top outer joint is for a 1¿ immersion thermometer while the 10/30 joints are for a 2¿ immersion thermometer. The 24/40 joint size uses 4mm glass or PTFE stopcocks, and are positioned to permit changing of receiver without interruption of the distillation process. The 14/20 size uses 2mm bore glass or PTFE stopcocks.
Catalog No. CG124708 Supplier Chemglass Life Sciences Supplier No. CG124708
Distilling Head, Vigreux, Jacketed, 10/18 Thermometer Joint,14/20 Joint Size, 2mm Glass Stopcock, Approx. 255mm Height x 310mm Width. Jacketed fractionation distilling head for use under vacuum. The 10/18 top outer joint is for a 1¿ immersion thermometer while the 10/30 joints are for a 2¿ immersion thermometer. The 24/40 joint size uses 4mm glass or PTFE stopcocks, and are positioned to permit changing of receiver without interruption of the distillation process. The 14/20 size uses 2mm bore glass or PTFE stopcocks.