Learn More
Chemglass Life Sciences Tube Only, Distilling Receiver, Graduated, 14/20 Joint, 12mL, 135mm Height
SDP
Catalog No. CG127801
Encompass_Preferred
Tube Only, Distilling Receiver, Graduated, 14/20 Joint, 12mL, 135mm Height. Centrifuge tube shaped receiver with an upper standard taper joint has two glass hooks 180¿ apart. Graduated from 0 to 2mL in 0.1mL subdivisions and from 2 to 12 or 15mL in 0.2mL.
Catalog No. CG127801 Supplier Chemglass Life Sciences Supplier No. CG127801
Tube Only, Distilling Receiver, Graduated, 14/20 Joint, 12mL, 135mm Height. Centrifuge tube shaped receiver with an upper standard taper joint has two glass hooks 180¿ apart. Graduated from 0 to 2mL in 0.1mL subdivisions and from 2 to 12 or 15mL in 0.2mL.