Learn More
Chemglass Life Sciences Distilling Receiver, 20mL, Moisture Test, Dean-Stark, 10/30 Thermometer Joint, Modified, 24/40 Joint Size, 2mm PTFE Stopcock, Approx. 400mm Height x 130mm Width
SDP
Catalog No. CG126506
Encompass_Preferred
Distilling Receiver, 20mL, Moisture Test, Dean-Stark, 10/30 Thermometer Joint, Modified, 24/40 Joint Size, 2mm PTFE Stopcock, Approx. 400mm Height x 130mm Width. Modified Dean-Stark receiver designed for solvents that are either lighter or heavier than water. The lower three-way PTFE stopcock allows for the withdraw or return of the distillate during the distillation process. With a 10/30 outer joint on the top of the vapor tube for a 3” immersion thermometer. Graduations are the same as CG-1261.
Catalog No. CG126506 Supplier Chemglass Life Sciences Supplier No. CG126506
Distilling Receiver, 20mL, Moisture Test, Dean-Stark, 10/30 Thermometer Joint, Modified, 24/40 Joint Size, 2mm PTFE Stopcock, Approx. 400mm Height x 130mm Width. Modified Dean-Stark receiver designed for solvents that are either lighter or heavier than water. The lower three-way PTFE stopcock allows for the withdraw or return of the distillate during the distillation process. With a 10/30 outer joint on the top of the vapor tube for a 3” immersion thermometer. Graduations are the same as CG-1261.