Learn More
Chemglass Life Sciences Distilling Receiver, 10mL, Moisture Test, Barrett, 24/40 Joint Size, 2mm PTFE Stopcock, Approx. 275mm Height x 95mm Width
SDP
Catalog No. 50873057
Encompass_Preferred
Distilling Receiver, 10mL, Moisture Test, Barrett, 24/40 Joint Size, 2mm PTFE Stopcock, Approx. 275mm Height x 95mm Width. Similar to CG-1261 but with the addition of an overflow tube between the uptake tube and the graduated receiver.
Catalog No. 50-873-057 Supplier Chemglass Life Sciences Supplier No. CG126310
Distilling Receiver, 10mL, Moisture Test, Barrett, 24/40 Joint Size, 2mm PTFE Stopcock, Approx. 275mm Height x 95mm Width. Similar to CG-1261 but with the addition of an overflow tube between the uptake tube and the graduated receiver.