Learn More
Chemglass Life Sciences Distilling Receiver, 15mL, 14/20, Complete
SDP
Catalog No. CG127614
Encompass_Preferred
Distilling Receiver, 15mL, 14/20, Complete. Cow type design has three receivers 45º apart. Condensers with lower drip tip inner joints will protrude into the spherical section, permitting the distillate to fall directly into the receiving flasks. Distribution adapter has a serrated connection for attachment to vacuum source. Complete item supplied with distribution adapter, three flasks of the listed size and six stainless steel springs.
Catalog No. CG127614 Supplier Chemglass Life Sciences Supplier No. CG127614
Distilling Receiver, 15mL, 14/20, Complete. Cow type design has three receivers 45º apart. Condensers with lower drip tip inner joints will protrude into the spherical section, permitting the distillate to fall directly into the receiving flasks. Distribution adapter has a serrated connection for attachment to vacuum source. Complete item supplied with distribution adapter, three flasks of the listed size and six stainless steel springs.