Learn More
Chemglass Life Sciences Distilling Receiver, 12mL, 14/20, Complete, Approx. 175mm Height x 240mm Width
SDP
Catalog No. CG127514
Encompass_Preferred
Distilling Receiver, 12mL, 14/20, Complete, Approx. 175mm Height x 240mm Width. The 135º angle between the center line of the condenser and the receiver tubes allows the distillate to fall directly into the receiver tubes. This receiver can be used with short path style distilling heads. Complete receiver is supplied with distribution adapter, three 12mL receiver tubes, and six CG-110 stainless steel springs.
Catalog No. CG127514 Supplier Chemglass Life Sciences Supplier No. CG127514
Distilling Receiver, 12mL, 14/20, Complete, Approx. 175mm Height x 240mm Width. The 135º angle between the center line of the condenser and the receiver tubes allows the distillate to fall directly into the receiver tubes. This receiver can be used with short path style distilling heads. Complete receiver is supplied with distribution adapter, three 12mL receiver tubes, and six CG-110 stainless steel springs.