Learn More
Chemglass Life Sciences Distilling Receiver, 20mL, Moisture Test, Barrett, 24/40 Joint Size, 2mm Glass Stopcock, Approx. 325mm Height x 95mm Width
SDP
Supplier:  Chemglass Life Sciences CG126306
Encompass_Preferred
Distilling Receiver, 20mL, Moisture Test, Barrett, 24/40 Joint Size, 2mm Glass Stopcock, Approx. 325mm Height x 95mm Width. Similar to CG-1261 but with the addition of an overflow tube between the uptake tube and the graduated receiver.
Catalog No. 50-873-056