Learn More
Chemglass Life Sciences Distilling Receiver, 25mL, Moisture Test, Dean-Stark, 24/40 Joint Size, Approx. 330mm Height x 90mm Width
SDP
Supplier:  Chemglass Life Sciences CG125904
Encompass_Preferred
Distilling Receiver, 25mL, Moisture Test, Dean-Stark, 24/40 Joint Size, Approx. 330mm Height x 90mm Width. Similar to CG-1258 but with the addition of an overflow tube between the uptake tube and the graduated receiver.
Catalog No. CG125904