Learn More
Chemglass Life Sciences Distilling Receiver, 25mL, Moisture Test, Dean-Stark, 14/20 Joint Size, Approx. 330mm Height x 80mm Width
SDP
Catalog No. 50873052
Encompass_Preferred
Distilling Receiver, 25mL, Moisture Test, Dean-Stark, 14/20 Joint Size, Approx. 330mm Height x 80mm Width. Dean-Stark receiver used for determining the water content of various materials. Can be used with ASTM Method D-95 along with similar ASTM methods. The 10mL size has subdivisions of 0.1 from 0 to 2mL, and 0.2 from 2 to 10mL. The 25mL size has subdivisions of 0.1 from 0 to 2mL and 0.2 from 2 to 25mL.
Catalog No. 50-873-052 Supplier Chemglass Life Sciences Supplier No. CG125815
Distilling Receiver, 25mL, Moisture Test, Dean-Stark, 14/20 Joint Size, Approx. 330mm Height x 80mm Width. Dean-Stark receiver used for determining the water content of various materials. Can be used with ASTM Method D-95 along with similar ASTM methods. The 10mL size has subdivisions of 0.1 from 0 to 2mL, and 0.2 from 2 to 10mL. The 25mL size has subdivisions of 0.1 from 0 to 2mL and 0.2 from 2 to 25mL.