Learn More
Chemglass Life Sciences Distilling Receiver, 10mL, Moisture Test, Merlic, 14/20 Joint Size, 2mm PTFE Stopcock, Approx. 275mm Height x 75mm Width
SDP
Catalog No. CG126404
Encompass_Preferred
Distilling Receiver, 10mL, Moisture Test, Merlic, 14/20 Joint Size, 2mm PTFE Stopcock, Approx. 275mm Height x 75mm Width. Modified Dean-Stark trap allows for more efficient cooling and separation of the azeotrope during distillation. Graduated from 0 to 10ml in 0.2mL subdivisions.Design suggested by Dr. Craig Merlic, University of California, Los Angeles, California.
Catalog No. CG126404 Supplier Chemglass Life Sciences Supplier No. CG126404
Distilling Receiver, 10mL, Moisture Test, Merlic, 14/20 Joint Size, 2mm PTFE Stopcock, Approx. 275mm Height x 75mm Width. Modified Dean-Stark trap allows for more efficient cooling and separation of the azeotrope during distillation. Graduated from 0 to 10ml in 0.2mL subdivisions.Design suggested by Dr. Craig Merlic, University of California, Los Angeles, California.