Learn More
Chemglass Life Sciences Distilling Receiver, 3mL, 14/20 Joint, Graduated, 210mm height x 90mm width
SDP
Catalog No. CG127714
Encompass_Preferred
Distilling Receiver, 3mL, 14/20 Joint, Graduated, 210mm height x 90mm width. Cow type receiver has a single standard taper outer joint at the top. The four lower graduated receiver tubes can be accessed via a syringe for product recovery. Tubes are graduated from 0 to 3mL in 0.2mL subdivisions.
Catalog No. CG127714 Supplier Chemglass Life Sciences Supplier No. CG127714
Distilling Receiver, 3mL, 14/20 Joint, Graduated, 210mm height x 90mm width. Cow type receiver has a single standard taper outer joint at the top. The four lower graduated receiver tubes can be accessed via a syringe for product recovery. Tubes are graduated from 0 to 3mL in 0.2mL subdivisions.