Learn More
Ace Glass, Inc. Distilling unit, 14/20 PTFE clad joints, complete with (4) 5mLflasks
SDP
Catalog No. 50976762
Encompass_Preferred
Distilling unit used for obtaining separate fractions while maintaining constant pressure. Unit has 14/20 PTFE-clad joints, and is supplied with (4) 5mL flasks.
Catalog No. 50-976-762 Supplier Ace Glass, Inc. Supplier No. 940025
Distilling unit used for obtaining separate fractions while maintaining constant pressure. Unit has 14/20 PTFE-clad joints, and is supplied with (4) 5mL flasks.