Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pain Care Labs DistrACTION™ Cards 4-Pack
SDP

Catalog No. 22450106
Change view
Click to view available options
Quantity:
4 Pk.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
22-450-106 4 Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 22-450-106 Supplier Pain Care Labs Supplier No. DTCSET
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Pain Care Labs DistrACTION™ Cards 4-Pack contains everything you need to relieve the pain and stress of any prickly situation, plus on-the-go entertainment.

  • Latex-free
  • Kit Includes: 1 Buzzy™ Mini Personal (Bee-Striped), 2 Universal Soft Wings, 1 Silicone Comfort Strap, 2 AAA Batteries (preinstalled in unit), 4 Needle Phobia Education Cards, 3 Sets of DistrACTION™ cards (assorted), 1 Instruction Manual, 6 Bravery Badge Stickers
  • Relieves the pain from minor sharp aches and pains, including needles, splinters, and stings.
  • Each Buzzy kit includes a Buzzy, instructions, a set of Universal Personal Ice Wings, a hands-free strap and batteries.
  • Distraction can reduce pain by up to 50%, often better than medical pain relief.
  • Five brightly colored cards on a snap ring, with age-specific questions and finding tasks for all ages.

Specifications

Quantity 4 Pk.
Product Type Distraction Cards
Color Assorted
For Use With (Application) Needle procedures
Material Teslin
Dimensions (L x W x H) 2 x 5.5 x 9 cm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.