Learn More
Synthware™ Vacuum Adapter for Distribution Receiver, Vertical
GREENER_CHOICESDP
Catalog No. 31501051 Shop All Kemtech Products
Change view
Click to view available options
Includes:
Hose Connection and Vertical Orientation
Standard Taper Joint:
24/40
Catalog No. Includes Standard Taper Joint
31-501-051 Hose Connection and Vertical Orientation 24/40
1 options
Catalog No. 31-501-051 Supplier Synthware™ Supplier No. A592440
Add to Cart
Add to Cart
Rotating distillation receiver allows four fractions to be collected. Vacuum adapter has a vertical orientation.
Specifications
Includes Hose Connection and Vertical Orientation
For Use With (Application) Organic Synthesis
Type Distribution Adapter
Material Borosilicate Glass
For Use With (Equipment) Distillation Set
Quantity 1 Ea.
Standard Taper Joint 24/40
Joint Type Ground Glass
Autoclavable Autoclavable