Learn More
Bellco Glass, Inc. Bottle Rinser
SDP
Catalog No. 501946083
Encompass_Preferred
The bottle rinser rinses any bottle over 75mm inside height from 10-102mm. Connects to a standard laboratory water tap with a max of 80 psig. Pressurized water reaches every corner with a unique swirling action. Includes 1m length of 3 ply high pressure hose.. MADE IN THE USA
Catalog No. 50-194-6083 Supplier Bellco Glass, Inc. Supplier No. 773188290
The bottle rinser rinses any bottle over 75mm inside height from 10-102mm. Connects to a standard laboratory water tap with a max of 80 psig. Pressurized water reaches every corner with a unique swirling action. Includes 1m length of 3 ply high pressure hose.. MADE IN THE USA