Learn More
Heidolph™ Distributor, to Expansion Vessel for Laborota 20, Hei-VAP Industrial Rotary Evaporator Glassware
SDP
Catalog No. 13900966 Shop All Heidolph North America Products
Change view
Click to view available options
For Use With (Equipment):
Laborota 20, Hei-VAP Industrial Rotary Evaporator
Catalog No. For Use With (Equipment)
13-900-966 Laborota 20, Hei-VAP Industrial Rotary Evaporator
1 options
Catalog No. 13-900-966 Supplier Heidolph™ Supplier No. 036304420
Add to Cart
Add to Cart
Distributor, to expansion vessel connects drive head to expansion vessel.
Included with Laborota 20, Hei-VAP Industrial Rotary Evaporator Glassware set A, AC, A1, A1C, A2, and A2C
Specifications
Type Glassware
For Use With (Equipment) Laborota 20, Hei-VAP Industrial Rotary Evaporator
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov