Learn More
Ace Glass, Inc. Distribution receiver only, short path, 14/20 joints, hose connection for 5/16 inch ID tubing
SDP
Supplier:  Ace Glass, Inc. 940308
Encompass_Preferred
Distribution receiver only that connects to three 25mL flasks in same plane, and rotate about the condenser axis. Hose connection is parallel to and above condenser axis. Three clips are supplied for holding flasks to condenser. All joints are 14/20. Hose connection is size A.
Catalog No. 50-976-764