Learn More
Cayman Chemical Disuccinimidyl Glutarate, Purity: ≥95%, CAS: 79642-50-5, 25 mg
SDP
Catalog No. 502982914
Encompass_Preferred
A homobifunctional crosslinking agent that contains amine-reactive NHS ester groups at each end; can be combined with formaldehyde fixation to improve the detection of specific protein-DNA complexes
Catalog No. 50-298-2914 Supplier Cayman Chemical Supplier No. 2064625 MG
A homobifunctional crosslinking agent that contains amine-reactive NHS ester groups at each end; can be combined with formaldehyde fixation to improve the detection of specific protein-DNA complexes