Learn More
Cayman Chemical DIsuccInImIdyl Glutarate 50mg
SDP
Catalog No. 502984504
Encompass_Preferred
A homobifunctional crosslinking agent that contains amine-reactive NHS ester groups at EAch end; can be combined with formaldehyde fixation to improve the detection of specific protein-DNA complexes
Catalog No. 50-298-4504 Supplier Cayman Chemical Supplier No. 2064650 MG
A homobifunctional crosslinking agent that contains amine-reactive NHS ester groups at EAch end; can be combined with formaldehyde fixation to improve the detection of specific protein-DNA complexes