Learn More
Medchemexpress LLC Disuccinimidyl glutarate | 79642-50-5 | 98.0% | 326.26 | 500 MG
SDP
Catalog No. 5000436790
Encompass_Preferred
DSG Crosslinker is an amine-reactive and noncleavable protein crosslinker that enables stable protein-protein, peptide, and biomolecule conjugation.
  • Amine-reactive
  • Noncleavable protein crosslinker
  • Enables stable protein-protein, peptide, and biomolecule conjugation
Catalog No. 50-004-36790 Supplier Medchemexpress LLC Supplier No. HY114697500MG
DSG Crosslinker is an amine-reactive and noncleavable protein crosslinker that enables stable protein-protein, peptide, and biomolecule conjugation.
  • Amine-reactive
  • Noncleavable protein crosslinker
  • Enables stable protein-protein, peptide, and biomolecule conjugation