Learn More
Medchemexpress LLC Disuccinimidyl sebacate | 23024-29-5 | 97.0% | 100 G
SDP
Catalog No. 5000234233
Encompass_Preferred
Disuccinimidyl sebacate is a homobifunctional, amine-reactive N-hydroxysuccinimide (NHS) ester cross-linker used to form stable amide bonds between primary amines in proteins and peptides. It is commonly used to prepare intermolecular cross-linked peptides and to stabilize protein-protein interactions for biochemical and structural studies.
  • Homobifunctional NHS ester reactive toward primary amines.
  • Enables intermolecular cross-linking of peptides and proteins.
  • Useful for stabilizing protein-protein interactions for analysis.
  • High purity (97.0%), suitable for biochemical assays.
  • Available in multiple pack sizes, including 100 g.
  • Store powder at -20°C; in solvent store at -80°C for long-term.
Catalog No. 50-002-34233 Supplier Medchemexpress LLC Supplier No. HY119523100G
Disuccinimidyl sebacate is a homobifunctional, amine-reactive N-hydroxysuccinimide (NHS) ester cross-linker used to form stable amide bonds between primary amines in proteins and peptides. It is commonly used to prepare intermolecular cross-linked peptides and to stabilize protein-protein interactions for biochemical and structural studies.
  • Homobifunctional NHS ester reactive toward primary amines.
  • Enables intermolecular cross-linking of peptides and proteins.
  • Useful for stabilizing protein-protein interactions for analysis.
  • High purity (97.0%), suitable for biochemical assays.
  • Available in multiple pack sizes, including 100 g.
  • Store powder at -20°C; in solvent store at -80°C for long-term.