Learn More
Cayman Chemical Disuccinimidyl Suberate, Purity: ≥98%, CAS: 68528-80-3, 250 mg
SDP
Catalog No. 502953065
Encompass_Preferred
A bifunctional cross-linking reagent reactive toward primary amines; is membrane-permable and commonly used to cross-link proteins in intact cell
Catalog No. 50-295-3065 Supplier Cayman Chemical Supplier No. 13008250 MG
A bifunctional cross-linking reagent reactive toward primary amines; is membrane-permable and commonly used to cross-link proteins in intact cell