Learn More
Abcam Disuccinimidyl suberate, cross-linking reagent, 1G
Catalog No. 502706727
Encompass_Preferred
MW 368.34 Da, Purity >99%. Homobifunctional primary amine reactive cross-linker. Contains an 8 atom linker (11.4 Å) and can cross-link distances up to ~25 Å due to protein flexibility. Produces no toxic side products.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-6727 Supplier Abcam Supplier No. AB1412741G
MW 368.34 Da, Purity >99%. Homobifunctional primary amine reactive cross-linker. Contains an 8 atom linker (11.4 Å) and can cross-link distances up to ~25 Å due to protein flexibility. Produces no toxic side products.
The product is subject to the following: Abcam Restricted Use Statement