Learn More
Cayman Chemical DIsuccInImIdyl SulfoxIde 100mg
SDP
Catalog No. 502895222
Encompass_Preferred
A protein cross-linking probe designed to be used with MS; contains two symmetric collision-induced dissociation-cleavable sites that allow effective identification of DSSO-cross-linked peptides based on their distinct fragmentation patterns
Catalog No. 50-289-5222 Supplier Cayman Chemical Supplier No. 9002863100 MG
A protein cross-linking probe designed to be used with MS; contains two symmetric collision-induced dissociation-cleavable sites that allow effective identification of DSSO-cross-linked peptides based on their distinct fragmentation patterns