Learn More
Cayman Chemical Disuccinimidyl Sulfoxide, Purity: ≥95%, CAS: 1351828-03-9, 50 mg
SDP
Catalog No. 502895733
Encompass_Preferred
A protein cross-linking probe designed to be used with MS; contains two symmetric collision-induced dissociation-cleavable sites that allow effective identification of DSSO-cross-linked peptides based on their distinct fragmentation patterns
Catalog No. 50-289-5733 Supplier Cayman Chemical Supplier No. 900286350 MG
A protein cross-linking probe designed to be used with MS; contains two symmetric collision-induced dissociation-cleavable sites that allow effective identification of DSSO-cross-linked peptides based on their distinct fragmentation patterns