Learn More
Medchemexpress LLC HY-B0240 500mg Medchemexpress, Disulfiram CAS:97-77-8 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYB0240500MG
Encompass_Preferred
Medchemexpress, HY-B0240 500mg Disulfiram CAS:97-77-8 Disulfiram (Tetraethylthiuram disulfide) is a specific inhibitor of aldehyde-dehydrogenase (ALDH1), used for the treatment of chronic alcoholism by producing an acute sensitivity to alcohol. Disulfiram inhibits gasdermin D (GSDMD) pore formation in liposomes and inflammasome-mediated pyroptosis and IL-1β secretion in human and mouse cells[1-4]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-9394