Learn More
Cayman Chemical Disulfiram, 97-77-8, 50 g
SDP
Supplier:  Cayman Chemical 1530350
Encompass_Preferred
Molecular Formula C10H20N2S4, Purity ≥95%, Formula Weight 296.5, Synonyms: N,N,N',N'-tetraethyl-thioperoxydicarbonic diamide, NSC 25953, Tetraethylthiuram disulfide; A copper and zinc chelator and an irreversible inhibitor of aldehyde dehydrogenase (IC50 = 0.1 mM) that has been indicated for the treatment of alcohol dependence; targets the ubiquitin-proteasome pathway, induces ROS, activates JNK and p38 pathways, and inhibits NF-B activity in cancer cells.
Catalog No. NC1221224