Learn More
Selleck Chemical LLC Disulfiram-50mg
SDP
Catalog No. 502389743
Encompass_Preferred
Disulfiram is a specific inhibitor of aldehyde-dehydrogenase (ALDH) with IC50 of 0.15 μM and 1.45 μM for hALDH1 and hALDH2, respectively. Disulfiram is used for the treatment of chronic alcoholism by producing an acute sensitivity to alcohol. Disulfiram induces apoptosis. Disulfiram is also an inhibitor of pore formation by gasdermin D (GSDMD).
Catalog No. 50-238-9743 Supplier Selleck Chemical LLC Supplier No. S168050MG
Add to Cart
Add to Cart
Disulfiram is a specific inhibitor of aldehyde-dehydrogenase (ALDH) with IC50 of 0.15 μM and 1.45 μM for hALDH1 and hALDH2, respectively. Disulfiram is used for the treatment of chronic alcoholism by producing an acute sensitivity to alcohol. Disulfiram induces apoptosis. Disulfiram is also an inhibitor of pore formation by gasdermin D (GSDMD).