Learn More
Medchemexpress LLC HY-B0240 5g , Disulfiram Tetraethylthiuram disulfide;TETD CAS:97-77-8 Purity:98%
SDP
Catalog No. 501492203
Encompass_Preferred
Medchemexpress, HY-B0240 5g Disulfiram Tetraethylthiuram disulfide;TETD CAS:97-77-8 Formula:C10H20N2S4 Purity:98% Disulfiram is a specific inhibitor of  aldehyde-dehydrogenase (ALDH1) , used for the treatment of chronic alcoholism by producing an acute sensitivity to alcohol. Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-149-2203 Supplier Medchemexpress LLC Supplier No. HYB02405G
Medchemexpress, HY-B0240 5g Disulfiram Tetraethylthiuram disulfide;TETD CAS:97-77-8 Formula:C10H20N2S4 Purity:98% Disulfiram is a specific inhibitor of  aldehyde-dehydrogenase (ALDH1) , used for the treatment of chronic alcoholism by producing an acute sensitivity to alcohol. Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.