Learn More
Cayman Chemical Disulfiram-d20, Purity: ≥99% deuterated forms (d1-d20), CAS: 1216403-88-1, 1 mg
SDP
Catalog No. 502939181
Encompass_Preferred
An internal standard for the quantification of disulfiram by GC- or LC-MS
Catalog No. 50-293-9181 Supplier Cayman Chemical Supplier No. 285361 MG
An internal standard for the quantification of disulfiram by GC- or LC-MS