Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC DiSulfo-Cy5 alkyne | 1617497-19-4 | C35H40N3NaO7S2 | 5 MG
SDP

Catalog No. 5000372397
Encompass_Preferred

DiSulfo-Cy5 alkyne is a fluorescent dye utilized as a click chemistry reagent. This compound is provided as a solid with high purity, suitable for various research applications. It is important to store it properly to maintain its stability and effectiveness.

  • Molecular weight: 701.83
  • Purity: 98.97%
  • Appearance: Solid
  • Color: Brown to dark brown
  • In vitro solubility: DMSO 50 mg/mL (71.24 mM)
  • Storage conditions: -20°C, sealed, away from moisture and light
  • Storage in solvent: -80°C for 6 months; -20°C for 1 month

Catalog No. 50-003-72397 Supplier Medchemexpress LLC Supplier No. HY1517015MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

DiSulfo-Cy5 alkyne is a fluorescent dye utilized as a click chemistry reagent. This compound is provided as a solid with high purity, suitable for various research applications. It is important to store it properly to maintain its stability and effectiveness.

  • Molecular weight: 701.83
  • Purity: 98.97%
  • Appearance: Solid
  • Color: Brown to dark brown
  • In vitro solubility: DMSO 50 mg/mL (71.24 mM)
  • Storage conditions: -20°C, sealed, away from moisture and light
  • Storage in solvent: -80°C for 6 months; -20°C for 1 month

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.