Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Disulfo-ICG carboxylic acid | 2757507-18-7 | 95.4% | 935.04 | C45H48N2Na2O11S3 | 10 MG
SDP

Catalog No. 5000296956
Encompass_Preferred

Disulfo-ICG carboxylic acid is a carboxyl-functionalized derivative of indocyanine green (ICG), supplied as a near-infrared (NIR) fluorescent probe for research applications such as imaging and labeling. It is provided as a green solid with high purity for conjugation workflows.

  • Near-infrared (NIR) fluorescent dye derived from indocyanine green (ICG).
  • Contains a carboxylic acid functional group for bioconjugation.
  • High purity suitable for research applications.
  • Molecular weight approximately 935.04 g/mol for mass-based calculations.
  • Available in small, lab-scale pack sizes for experimental use.

Catalog No. 50-002-96956 Supplier Medchemexpress LLC Supplier No. HYD225010MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Disulfo-ICG carboxylic acid is a carboxyl-functionalized derivative of indocyanine green (ICG), supplied as a near-infrared (NIR) fluorescent probe for research applications such as imaging and labeling. It is provided as a green solid with high purity for conjugation workflows.

  • Near-infrared (NIR) fluorescent dye derived from indocyanine green (ICG).
  • Contains a carboxylic acid functional group for bioconjugation.
  • High purity suitable for research applications.
  • Molecular weight approximately 935.04 g/mol for mass-based calculations.
  • Available in small, lab-scale pack sizes for experimental use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.