Learn More
Crescent Chemical Co Inc DISULFOTON-SULFONE 50 MG
SDP
Supplier:  Crescent Chemical Co Inc C12985000
Encompass_Preferred
Disulfoton-sulfone Insecticide (Metabolite neat) CAS 2497-06-5 Molecular Weight - 306.4028 Storage - -18°C Catalog Num - DRE-CA12985000 pesticide
Catalog No. 50-776-91