Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MSC ZEP Solid Warewash Detergent, Unscented, 32 lb

Catalog No. 502817790
Encompass

ZEP ProVisions Solid Warewash Detergent SMS is an automatic warewash detergent specifically formulated for food and beverage facility maintenance. This white, solid detergent exhibits a characteristic odor, high alkalinity with a pH greater than 11.5 in aqueous solution, and is completely water-soluble.

  • Formulated for food and beverage facility maintenance
  • Automatic dishwashing detergent type
  • High alkalinity with pH greater than 11.5 (aqueous solution)
  • Density greater than 1.5 g/cm3
  • Completely soluble in water and acetone
  • Unscented composition
  • Contains sodium metasilicate (disodium salt) at 30-60% concentration

Catalog No. 50-281-7790 Supplier MSC Supplier No. 17596198
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

ZEP ProVisions Solid Warewash Detergent SMS is an automatic warewash detergent specifically formulated for food and beverage facility maintenance. This white, solid detergent exhibits a characteristic odor, high alkalinity with a pH greater than 11.5 in aqueous solution, and is completely water-soluble.

  • Formulated for food and beverage facility maintenance
  • Automatic dishwashing detergent type
  • High alkalinity with pH greater than 11.5 (aqueous solution)
  • Density greater than 1.5 g/cm3
  • Completely soluble in water and acetone
  • Unscented composition
  • Contains sodium metasilicate (disodium salt) at 30-60% concentration

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.