Learn More
Crescent Chemical Co Inc DITHIBIS(2-NITROBENZOIC ACID)
SDP
Catalog No. 50247533
Encompass_Preferred
Dithiobis(2-nitrobenzoic acid) research grade For qualitative and quantitative determination of sulfhydryl groups. Size - 5G Storage Conditions - +15 °C TO +30 °C Catalog Number - 20735.02 CAS 69-78-3
Catalog No. 50-247-533 Supplier Crescent Chemical Co Inc Supplier No. SE20735.02
Dithiobis(2-nitrobenzoic acid) research grade For qualitative and quantitative determination of sulfhydryl groups. Size - 5G Storage Conditions - +15 °C TO +30 °C Catalog Number - 20735.02 CAS 69-78-3