Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dithieno[3,2-c:2',3'-e]oxepine-4,6-dione 98.0+%, TCI America™
SDP

Catalog No. D49721G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D49721G 1 g
D49725G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D49721G Supplier TCI America Supplier No. D49721G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 1043023-52-4
Molecular Formula C10H4O3S2
Molecular Weight (g/mol) 236.259
InChI Key UGFYLGZFYCTMFU-UHFFFAOYSA-N
PubChem CID 89807128
IUPAC Name dithieno[2,3-a
SMILES C1=CSC2=C1C(=O)OC(=O)C3=C2SC=C3

Specifications

Melting Point 263°C
Quantity 1 g
Formula Weight 236.26
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material Dithieno[3,2-c:2′,3′-e]oxepine-4,6-dione
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.