Learn More
Medchemexpress LLC Dithiodipropionic acid | 1119-62-6 | MFCD00002780 | 99.0% | 210.26 g·mol⁻¹ | C6H10O4S2 | 25 G
SDP
Catalog No. 5000303509
Encompass_Preferred
Dithiodipropionic acid is a disulfide-containing dicarboxylic acid (CAS 1119-62-6) commonly used as a building block and reagent in organic synthesis to introduce cleavable disulfide linkages and to functionalize materials. It is supplied as a high-purity solid suitable for research applications.
  • High purity (99.0%), suitable for research-grade applications.
  • Disulfide-containing dicarboxylic acid for introducing cleavable linkages.
  • Available in multiple pack sizes including 25 g for scalability.
  • Molecular formula C6H10O4S2 and molecular weight 210.26 g·mol⁻¹ for precise stoichiometry.
  • Commonly used in organic synthesis, nanoparticle functionalization, and material modification.
Catalog No. 50-003-03509 Supplier Medchemexpress LLC Supplier No. 25GHYW014395
Dithiodipropionic acid is a disulfide-containing dicarboxylic acid (CAS 1119-62-6) commonly used as a building block and reagent in organic synthesis to introduce cleavable disulfide linkages and to functionalize materials. It is supplied as a high-purity solid suitable for research applications.
  • High purity (99.0%), suitable for research-grade applications.
  • Disulfide-containing dicarboxylic acid for introducing cleavable linkages.
  • Available in multiple pack sizes including 25 g for scalability.
  • Molecular formula C6H10O4S2 and molecular weight 210.26 g·mol⁻¹ for precise stoichiometry.
  • Commonly used in organic synthesis, nanoparticle functionalization, and material modification.