Learn More
Research Products International Corp DTE [Dithioerythreitol], 5 Grams
SDP
Catalog No. 50213281
Encompass_Preferred
Protective reagent for maintaining -SH groups in reduced stated.
Hygroscopic.
Catalog No. 50-213-281 Supplier Research Products International Corp Supplier No. D200405.0
Protective reagent for maintaining -SH groups in reduced stated.
Hygroscopic.