Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dithioerythritol, MP Biomedicals™

Catalog No. ICN10059880 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
25 g
100 g
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN10059880 100 g
ICN10059801 1 g
ICN10059805 5 g
ICN10059810 10 g
ICN10059825 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. ICN10059880 Supplier MP Biomedicals, Inc Supplier No. 0210059880
Add to Cart
Add to Cart

Chemical Identifiers

CAS 6892-68-8
Molecular Formula C4H10O2S2
Molecular Weight (g/mol) 154.24
MDL Number MFCD00063750
InChI Key VHJLVAABSRFDPM-ZXZARUISSA-N
Synonym dithioerythritol, 1,4-dithioerythritol, dte, 2r,3s-1,4-dimercaptobutane-2,3-diol, erythro-1,4-dimercapto-2,3-butanediol, 2,3-butanediol, 1,4-dimercapto-, 2r,3s-rel, 1,4-dithioerythritol dte, 2r*,3s*-1,4-dimercapto-2,3-butanediol, erythro-2,3-dihydroxy-1,4-butanedithiol, 2r,3s-1,4-disulfanylbutane-2,3-diol
PubChem CID 439352
ChEBI CHEBI:17456
SMILES O[C@@H](CS)[C@H](O)CS

Specifications

Color White
Assay Percent Range >99%
Quantity 100 g
IUPAC Name (2R,3S)-1,4-bis(sulfanyl)butane-2,3-diol
Formula Weight 154.2
Physical Form Crystalline Powder
Chemical Name or Material Dithioerythritol
EINECSNumber 229-998-8
Recommended Storage Store at + 4°C, protect from light, store under nitrogen
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.