Learn More
Medchemexpress LLC Dithiooxamide | 79-40-3 | 99.9% | 120.20 | 100 G
SDP
Catalog No. 5000366459
Encompass_Preferred
Dithiooxamide (Rubeanic Acid) is a selective chelating agent and sulfur-containing analog of Oxamide.
It forms stable chelates with various metal ions, which can be separated, enriched, and detected by adsorption on solid adsorbents or electrode surfaces.
  • Selective chelating agent
  • Sulfur-containing analog of Oxamide
  • Forms stable chelates with metal ions (lead, iron, cadmium, manganese, aluminum)
  • Chelates can be separated, enriched, and detected by adsorption on solid adsorbents or electrode surfaces
Catalog No. 50-003-66459 Supplier Medchemexpress LLC Supplier No. HYW010488100G
Dithiooxamide (Rubeanic Acid) is a selective chelating agent and sulfur-containing analog of Oxamide.
It forms stable chelates with various metal ions, which can be separated, enriched, and detected by adsorption on solid adsorbents or electrode surfaces.
  • Selective chelating agent
  • Sulfur-containing analog of Oxamide
  • Forms stable chelates with metal ions (lead, iron, cadmium, manganese, aluminum)
  • Chelates can be separated, enriched, and detected by adsorption on solid adsorbents or electrode surfaces