Learn More
Medchemexpress LLC Dithiooxamide (Rubeanic Acid) | 79-40-3 | 99.9% | 120.20 | 25 G
SDP
Supplier:  Medchemexpress LLC 25GHYW010488
Encompass_Preferred
Dithiooxamide (Rubeanic Acid) is a selective chelating agent and a sulfur-containing analog of Oxamide. It forms stable chelates with various metal ions such as lead, iron, cadmium, manganese, and aluminum. These chelates can then be separated, enriched, and detected by adsorption on solid adsorbents or electrode surfaces.
  • Selective chelating agent
  • Sulfur-containing analog of Oxamide
  • Forms stable chelates with various metal ions
  • Chelates can be separated, enriched and detected
  • Available in various package sizes for research use
Catalog No. 50-004-51789