Learn More
Waters Corp Dithiothreitol, 16 mg, 8/pk, Reducing Agent
Catalog No. 502391851 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
Dithiothreitol (DTT) is a reducing agent designed to disrupt disulfide bonds in proteins, facilitating protein denaturing. It is supplied as 16 mg of DTT in powder form, housed in amber glass bottles with screw-top caps. Each pack contains 8 bottles, serving as a supplemental reagent for PeptideWorks Tryptic Protein Digestion Kits.
  • Disrupts protein disulfide bonds for denaturing.
  • 16 mg DTT in powder form per bottle.
  • 8 bottles per pack, for up to 96 samples.
  • Amber glass bottles with screw-top caps.
  • Supplemental reagent for PeptideWorks Tryptic Protein Digestion Kits.
  • Prepare by dissolving in 2.1 mL of 18.2 MΩ H2O.
  • Use freshly prepared solution within one day, then discard.
Catalog No. 50-239-1851 Supplier Waters Corp Supplier No. 186010603
Dithiothreitol (DTT) is a reducing agent designed to disrupt disulfide bonds in proteins, facilitating protein denaturing. It is supplied as 16 mg of DTT in powder form, housed in amber glass bottles with screw-top caps. Each pack contains 8 bottles, serving as a supplemental reagent for PeptideWorks Tryptic Protein Digestion Kits.
  • Disrupts protein disulfide bonds for denaturing.
  • 16 mg DTT in powder form per bottle.
  • 8 bottles per pack, for up to 96 samples.
  • Amber glass bottles with screw-top caps.
  • Supplemental reagent for PeptideWorks Tryptic Protein Digestion Kits.
  • Prepare by dissolving in 2.1 mL of 18.2 MΩ H2O.
  • Use freshly prepared solution within one day, then discard.