Learn More
AG Scientific Inc Dithiothreitol, DTT, 25 G
Catalog No. 502237028
Encompass_Preferred
Dithiothreitol (DTT), is a small-molecule redox reagent, known as Cleland's reagent, that forms a six-member ring with an internal disulfide bond. It has a redox potential of -0.33 V at pH 7. The reduction of typical disulfide bond proceeds by two sequential thiol-disulfide exchange reactions.DTT is effective in buffers to reduce protein disulfide bonds prior to SDS-PAGE. It can protect protein sulfhydryls and restore enzyme activity lost by oxidation.CAS Number: 3483-12-3Molecular Weight: 154.2Chemical Formula: C4H10O2S2Solubility: 10% solution in water. Clear and colorlessStorage Temperature: 2-8CResearch or further manufacturing use only, not for food or drug use.
Catalog No. 50-223-7028 Supplier AG Scientific Inc Supplier No. C102925GM
Dithiothreitol (DTT), is a small-molecule redox reagent, known as Cleland's reagent, that forms a six-member ring with an internal disulfide bond. It has a redox potential of -0.33 V at pH 7. The reduction of typical disulfide bond proceeds by two sequential thiol-disulfide exchange reactions.DTT is effective in buffers to reduce protein disulfide bonds prior to SDS-PAGE. It can protect protein sulfhydryls and restore enzyme activity lost by oxidation.CAS Number: 3483-12-3Molecular Weight: 154.2Chemical Formula: C4H10O2S2Solubility: 10% solution in water. Clear and colorlessStorage Temperature: 2-8CResearch or further manufacturing use only, not for food or drug use.