Learn More
Crescent Chemical Co Inc DITHIOTHREITOL MOL BIO GRADE
SDP
Supplier:  Crescent Chemical Co Inc SE39759.02
Encompass_Preferred
Dithiothreitol molecular biology grade DNase/RNase not detected. For quantitative reduction of disulfide groups (1). Oxidized DTT max. 0.5 %. Hygroscopic. Size - 5G Storage Conditions - +2 °C TO +8 °C Catalog Number - 39759.02 CAS 27565-
Catalog No. 50-248-442