Learn More
Medchemexpress LLC Dithranol (Anthralin) | 1143-38-0 | 95.3% | 50 MG
SDP
Catalog No. 5000405370
Encompass_Preferred
Dithranol (Anthralin) is an anthraquinone derivative with potent anti-psoriatic effects. It can inhibit DNA replication and repair.
  • Potent anti-psoriatic effects
  • Inhibits DNA replication and repair
  • Active against HEK293, HaCaT, PMNL, THP-1, U-937 cell lines
  • Used in clinical trials for Psoriasis Vulgaris
Catalog No. 50-004-05370 Supplier Medchemexpress LLC Supplier No. HYB073850MG
Dithranol (Anthralin) is an anthraquinone derivative with potent anti-psoriatic effects. It can inhibit DNA replication and repair.
  • Potent anti-psoriatic effects
  • Inhibits DNA replication and repair
  • Active against HEK293, HaCaT, PMNL, THP-1, U-937 cell lines
  • Used in clinical trials for Psoriasis Vulgaris