Learn More
Tecan 1000L Hanging Tips Fil
Catalog No. 50999849
Encompass_Preferred
Genuine Tecan 1000 L disposable automation tips for Freedom EVO Liquid Handling Arm™ (LiHa) or Fluent Flexible Channel Arm™ (FCA). Hanging tip format, filtered, conductive. 100 x 96-tip hanging racks, 9,600 tips/sales unit. LiHa Disposable Tips have been carefully designed and validated to work specifically with Liquid Handling Arms™ on all Tecan platforms. They are produced to stringent standards of quality and reproducibility, ensuring reliable liquid handling performance. Manufacturing is fully automated in a controlled environment, using state-of-the-art production processes, integrated quality control procedures and premium raw materials. During production, each tip undergoes in-line optical inspection using a CCD camera and image recognition software, checking all functional parts to ensure consistent pipetting accuracy and precision.
This item is not returnable. View return policy
Catalog No. 50-999-849 Supplier Tecan Supplier No. 10612513
Genuine Tecan 1000 L disposable automation tips for Freedom EVO Liquid Handling Arm™ (LiHa) or Fluent Flexible Channel Arm™ (FCA). Hanging tip format, filtered, conductive. 100 x 96-tip hanging racks, 9,600 tips/sales unit. LiHa Disposable Tips have been carefully designed and validated to work specifically with Liquid Handling Arms™ on all Tecan platforms. They are produced to stringent standards of quality and reproducibility, ensuring reliable liquid handling performance. Manufacturing is fully automated in a controlled environment, using state-of-the-art production processes, integrated quality control procedures and premium raw materials. During production, each tip undergoes in-line optical inspection using a CCD camera and image recognition software, checking all functional parts to ensure consistent pipetting accuracy and precision.