Learn More
Tecan 1000L Tips ANSI Fil
Catalog No. NC0283505
Encompass_Preferred
Genuine Tecan 1000 L disposable automation tips for Freedom EVO Liquid Handling Arm™ (LiHa) or Fluent Flexible Channel Arm™ (FCA). ANSI format, filtered, conductive. Tecan STERILE purity level - Sterile, tested & certified to be free from human DNA, Dnase, Rnase, pyrogens & endotoxins. 40 racks x 96-tip boxes, 3,840 tips/sales unit. LiHa Disposable Tips have been carefully designed & validated to work specifically with Liquid Handling Arms™ on all Tecan platforms. They are produced to stringent standards of quality & reproducibility, ensuring reliable liquid handling performance. Mfg is fully automated in a controlled environment, using state-of-the-art production processes, integrated quality control procedures & premium raw materials. During production, each tip undergoes in-line optical inspection using a CCD camera & image recognition software, checking all functional parts to ensure consistent pipetting accuracy & precision.
This item is not returnable. View return policy
Catalog No. NC0283505 Supplier Tecan Supplier No. 30057823
Genuine Tecan 1000 L disposable automation tips for Freedom EVO Liquid Handling Arm™ (LiHa) or Fluent Flexible Channel Arm™ (FCA). ANSI format, filtered, conductive. Tecan STERILE purity level - Sterile, tested & certified to be free from human DNA, Dnase, Rnase, pyrogens & endotoxins. 40 racks x 96-tip boxes, 3,840 tips/sales unit. LiHa Disposable Tips have been carefully designed & validated to work specifically with Liquid Handling Arms™ on all Tecan platforms. They are produced to stringent standards of quality & reproducibility, ensuring reliable liquid handling performance. Mfg is fully automated in a controlled environment, using state-of-the-art production processes, integrated quality control procedures & premium raw materials. During production, each tip undergoes in-line optical inspection using a CCD camera & image recognition software, checking all functional parts to ensure consistent pipetting accuracy & precision.