Learn More
Medchemexpress LLC HY-B1637 1g Medchemexpress, Ditiocarb sodium CAS:148-18-5 Purity:>98%
SDP
Catalog No. 501932982
Encompass_Preferred
Medchemexpress, HY-B1637 1g Ditiocarb sodium CAS:148-18-5 Ditiocarb sodium (Sodium diethyldithiocarbamate) is an accelerator of the rate of copper cementation. Sodium diethyldithiocarbamate reduces the incidence of HIV infection. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2982 Supplier Medchemexpress LLC Supplier No. HYB16371G
Medchemexpress, HY-B1637 1g Ditiocarb sodium CAS:148-18-5 Ditiocarb sodium (Sodium diethyldithiocarbamate) is an accelerator of the rate of copper cementation. Sodium diethyldithiocarbamate reduces the incidence of HIV infection. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.