Learn More
Medchemexpress LLC Ditiocarb sodium | 148-18-5 | 99.7% | 5 G
SDP
Catalog No. 5000413267
Encompass_Preferred
Ditiocarb sodium (Sodium diethyldithiocarbamate) is an orally active copper reagent used in the research of tumors, inflammatory, and immune-related diseases.
  • Exhibits antioxidation
  • Exhibits chelation
  • Exhibits anti-tumor effects
  • Exhibits immunomodulation
  • Exhibits anti-HIV properties
Catalog No. 50-004-13267 Supplier Medchemexpress LLC Supplier No. HYB16375G
Ditiocarb sodium (Sodium diethyldithiocarbamate) is an orally active copper reagent used in the research of tumors, inflammatory, and immune-related diseases.
  • Exhibits antioxidation
  • Exhibits chelation
  • Exhibits anti-tumor effects
  • Exhibits immunomodulation
  • Exhibits anti-HIV properties