Learn More
Medchemexpress LLC Ditolylguanidine | 97-39-2 | 99.66% | C15H17N3 | 500 MG
SDP
Catalog No. 5000354485
Encompass_Preferred
Ditolylguanidine (1,3-Di-o-tolylguanidine; DTG) is an agonist of sigma receptor (σ1/σ2 receptor) with Ki values of 69 and 21 nM for σ1 and σ2 receptors, respectively. It effectively inhibits the growth of small cell lung cancer cells and can be used for lung cancer research.
  • Agonist of sigma receptor (σ1/σ2 receptor)
  • Effectively inhibits the growth of small cell lung cancer cells
  • Suitable for lung cancer research
  • Inhibits N-(2-piperidinoethyl)4-iodobenzamide) (IPAB)
  • Affects micturition intervals and threshold pressure in anaesthetized rats
Catalog No. 50-003-54485 Supplier Medchemexpress LLC Supplier No. HY14218500MG
Ditolylguanidine (1,3-Di-o-tolylguanidine; DTG) is an agonist of sigma receptor (σ1/σ2 receptor) with Ki values of 69 and 21 nM for σ1 and σ2 receptors, respectively. It effectively inhibits the growth of small cell lung cancer cells and can be used for lung cancer research.
  • Agonist of sigma receptor (σ1/σ2 receptor)
  • Effectively inhibits the growth of small cell lung cancer cells
  • Suitable for lung cancer research
  • Inhibits N-(2-piperidinoethyl)4-iodobenzamide) (IPAB)
  • Affects micturition intervals and threshold pressure in anaesthetized rats